Graphic Controls Chart Paper, 8", -150 to +250 degree or -.15 to +.25, 7 day; 60/pk
Representative image only.
Graphic Controls Mfr # 01149731 – Item # EW-80013-32
Monitor with precision accuracy for quality control
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)8
  • Max Temperature (° F)250
  • Min Temperature (° F)-150
  • Max Temperature (° C)250
  • Min Temperature (° C)-150
  • DescriptionChart Paper, 8\", -150 to +250 degree or -.15 to +.25, 7 day; 60/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
  • 1 year warranty
More About this Item
Choose the paper to meet your application needs.