Great Plains Industries 125060-1 Turbine Flowmeter Output Module; Pulse Output
Representative image only.
Great Plains Industries Mfr # 125060-1 – Item # EW-05609-91
Optimize and enhance performance of your Masterflex flowmeter
  • A wide range of accessories are available to provide further functionality to your flowmeter
PRODUCT OPTIONS
$123.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • OutputPulse Output
  • DescriptionTurbine Flowmeter Output Module; Pulse Output
Key Features
  • A wide range of accessories are available to provide further functionality to your flowmeter
More About this Item
These accessories are ideal for the Masterflex® Heavy-Duty Turbine Flowmeters/Totalizers. Choode from the optional remote installation kit, replacement battery kits, output modules, and calibration containers to enhance performance of your flowmeters.
Module provides a digital open collector output for interfacing with compatible instrumentation.