Great Plains Industries 125100-1 Turbine Flowmeter Output Module: 4 to 20 mA
Representative image only.
Great Plains Industries Mfr # 125100-1 – Item # EW-05609-92
Optimize and enhance performance of your Masterflex flowmeter
  • A wide range of accessories are available to provide further functionality to your flowmeter
PRODUCT OPTIONS
$453.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Output (mA)4 to 20
  • Output (Voltage)5 to 20
  • Output4 to 20 mA or 5 to 20 volts
  • DescriptionTurbine Flowmeter Output Module: 4 to 20 mA
Key Features
  • A wide range of accessories are available to provide further functionality to your flowmeter
More About this Item
These accessories are ideal for the Masterflex® Heavy-Duty Turbine Flowmeters/Totalizers. Choode from the optional remote installation kit, replacement battery kits, output modules, and calibration containers to enhance performance of your flowmeters.
Module provides an output for interfacing with compatible instrumentation.