Great Plains Industries 1252 Vertical Pipe Mounting Kit for Turbine Flowmeters/Totalizers
Representative image only.
Great Plains Industries Mfr # 125260-01 – Item # EW-05611-90
Walk up and easily read your meter display
  • Allows horizontal reading of vertical meters
  • Easy to install
$62.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVertical Pipe Mounting Kit for Turbine Flowmeters/Totalizers
Key Features
  • Allows horizontal reading of vertical meters
  • Easy to install
More About this Item
Walk up and easily read your meter display without tilting your head. Easily convert your vertical meter to read horizontally.
90° Display kit allows for horizontal readout of vertical meters. Kit includes adapter, O-ring, screws, and foam spacers required for installation