Great Plains Industries Flowmeter for Fuel, Aluminum, 1" ISO Thread, 10 to 100 LPM
Representative image only.
Great Plains Industries Mfr # 113255-3 – Item # EW-32486-02
Small, powerful, and durable
  • Battery powered 4 digit display
  • Factory calibrated to water or fuel
  • Gallon or litre measurement units
  • Batch and cumulative totals
  • Ready to install
PRODUCT OPTIONS
$232.50USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5.0
  • Min Flow Rate (LPM)10
  • Max Flow Rate (LPM)100
  • Wetted MaterialsAluminum
  • Pipe Size1" ISO Thread
  • Fluid Typefuel and thin fluids
  • DescriptionFlowmeter for Fuel, Aluminum, 1" ISO Thread, 10 to 100 LPM
Key Features
  • Battery powered 4 digit display
  • Factory calibrated to water or fuel
  • Gallon or litre measurement units
  • Batch and cumulative totals
  • Ready to install
More About this Item
Small, powerful, microprocessor based totalizers powered by AAA batteries have large four-digit displays and mount at the nozzle for ultimate convenience. They are suited for flow ranges from 3 to 30 GPM. Nylon® models, rated to 150 PSIG, are factory calibrated for water. Aluminum models, rated to 300 PSIG are factory calibrated for fuels. Accuracy is ±5%. All mount with 1 inch female NPT fittings.