Grooving Tool for PFA Compression Fittings, 6 mm OD
Representative image only.
Item # EW-31321-49
Get a secure seal at higher pressures
  • Optimize performance of your PFA Compression Fittings
Free Certificate of Comliance
$385.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (mm)6
  • DescriptionGrooving Tool for PFA Compression Fittings, 6 mm OD
Certificate of Compliance
Key Features
  • Optimize performance of your PFA Compression Fittings
More About this Item
Improve the sealing capability of PTFE fittings. Fittings withstand pressures to 150 psi when a tool is used (40 psi without tool).