GT SCIEN TOGA® Puri Smart Filter for 81790-35, Type B
Representative image only.
Mfr # TOGA-S02D-AIF-B – Item # EW-81790-66
TOGA® filters are designed to enhance laboratory air quality and safeguard the health of researchers
  • Utilizes physical adsorption, chemical reaction, and neutralization for superior performance
  • No ozone or nitrogen oxide emission from side reactions compared to carbon filters
  • Fully utilizes filter passage area for effective pollutant removal
  • A Types are excellent for removing VOCs, alcohol, hydrocarbons, and odors
  • B Types offer efficient removal of general organic compounds, ozone, ammonia, and bad odors
  • C Types are great for removal of nitrogen and sulfur based oxides
  • 2 year warranty
PRODUCT OPTIONS
$1,584.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With81790-35 Toxic Gas Purifier
  • DescriptionTOGA® Puri Smart Filter for 81790-35, Type B
  • Warranty2 years
Key Features
  • Utilizes physical adsorption, chemical reaction, and neutralization for superior performance
  • No ozone or nitrogen oxide emission from side reactions compared to carbon filters
  • Fully utilizes filter passage area for effective pollutant removal
  • A Types are excellent for removing VOCs, alcohol, hydrocarbons, and odors
  • B Types offer efficient removal of general organic compounds, ozone, ammonia, and bad odors
  • C Types are great for removal of nitrogen and sulfur based oxides
  • 2 year warranty
More About this Item
The TOGA® filter signifies a paradigm shift in laboratory air quality management. In comparison to common carbon filters, the TOGA® filter's multifaceted approach—utilizing physical adsorption, chemical reaction, and neutralization—offers an array of advantages, including an extended filter replacement cycle, excellent purification of harmful gases, and minimized contamination risks.
With distinct types catering to various applications, from general organic compounds to specific chemical challenges, the TOGA® filter showcases its versatility. Rigorous performance tests conducted by reputable institutions attest to its consistently high removal efficiency, making it a reliable choice for laboratories seeking superior air quality solutions