GT SCIEN TOGA® Puri Smart, Portable Toxic Gas Purifier for Acid; 120V

Representative image only.
Mfr # TOGA-MAC1-AI – Item # EW-81790-39
The ultimate toxic gas purification solution for the laboratory
  • Three models to choose from
  • Standing or moveable versions
  • Easy to use intelligent touch panels
  • Efficient toxic gas intake
  • Puri Smart models add on toxic gas monitoring and AIoT integration for seamless automated control
  • 2 year warranty

PRODUCT OPTIONS

$8,268.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Blower TypeMotor with 5 level speed control
  • Exterior Width (in)18
  • Exterior Height (in)41
  • Exterior Depth (in)18
  • Power (VAC)120
  • Exhaust Volume (CFM)353
  • Shipping Weight (kg)70
  • DescriptionTOGA® Puri Smart, Portable Toxic Gas Purifier for Acid; 120V
  • Warranty2 years
Two year warranty

Key Features

  • Three models to choose from
  • Standing or moveable versions
  • Easy to use intelligent touch panels
  • Efficient toxic gas intake
  • Puri Smart models add on toxic gas monitoring and AIoT integration for seamless automated control
  • 2 year warranty

More About this Item

Standing models effectively and efficiently purify toxic gases that are generated during experimentation and from stored reagents. Portable models purify toxic gases at their source as they are generated, ensuring that researchers are not exposed to any harmful gases produced as a byproduct. Adopt both models to create and maintain a healthy indoor environment for researchers.

Puri SmartAI models add on toxic gas monitoring for automated operation. They will continuously monitor VOC levels within the laboratory. They can also seamlessly integrate with equipment for automated control using the SmartLab Sensor AI. A Puri SmartAI Pro may also accept a UPLA filter (standard HEPA) and a UVC lamp to eliminate bacteria and viruses.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat