Guardian Equipment Combination Safety Shower with Eyewash, Stainless Steel Bowl and Cover and ABS Plastic Shower Head
Representative image only.
Guardian Equipment Mfr # G1902BC – Item # EW-06767-45
Enhance safety with combination safety stations that feature a shower and eyewash bowl
  • Safety stations feature a 10” diameter shower head
  • GS-Plus™ spray heads provide flow control and filter to remove impurities from water
  • Easily visible ANSI-compliant identification sign
  • Furnished with orange polyethylene pipe covers for high visibility and corrosion resistance
  • Constructed with a stainless steel bowl and bowl cover and an orange ABS plastic shower head
  • 2 year warranty
PRODUCT OPTIONS
$1,578.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Flow Rate (GPM)3.6
  • Shower Flow Rate (GPM)20
  • Eyewash Fixture MaterialChrome-Plated Brass
  • Bowl MaterialStainless Steel
  • Shower Head MaterialABS Plastic
  • Supply Pipe MaterialSchedule 40 Galvanized Steel
  • MountFloor Mount
  • Outlet Connections1-1/4\" NPT(F)
  • Inlet Connection1-1/4\" NPT(F)
  • ComplianceANSI Z385.1-2009
  • DescriptionCombination Safety Shower with Eyewash, Stainless Steel Bowl and Cover and ABS Plastic Shower Head
  • Warranty2 Years
Two year warrantyCanadian Standards AssociationMade in the U.S.A.UL Listed.
Key Features
  • Safety stations feature a 10” diameter shower head
  • GS-Plus™ spray heads provide flow control and filter to remove impurities from water
  • Easily visible ANSI-compliant identification sign
  • Furnished with orange polyethylene pipe covers for high visibility and corrosion resistance
  • Constructed with a stainless steel bowl and bowl cover and an orange ABS plastic shower head
  • 2 year warranty
More About this Item
These Guardian Equipment combination safety stations feature a free-standing shower and an eyewash station for optimal safety. All stations feature a stay open ball valve in both the shower head and wash bowl. Furnished with a stainless steel actuating arm and pull rod for shower operation.
Choose from models with a stainless steel or an ABS plastic shower head or bowl construction, hand only or hand and foot operation, and standard or WideArea™ for improved access.