Guzzler 45 Hose for model 07090-11 (Tube length: 192)
Representative image only.
Guzzler Mfr # SG.H.-1 1/2 – Item # EW-06309-11
Optimize performance of your Guzzler® Diaphragm Hand Pump
  • Suitable for use with Guzzler® Portable Diaphragm Hand Pumps
PRODUCT OPTIONS
$66.85USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Tube Length, Inlet (in)192
  • Tube Length, Inlet (cm)488
  • DescriptionHose for model 07090-11 (Tube length: 192)
Key Features
  • Suitable for use with Guzzler® Portable Diaphragm Hand Pumps
More About this Item
These hoses are noncollapsing, wire-reinforced, and constructed of durable vinyl.
Model 06309-10 hose is compatible with Guzzler pumps 07090-09 and -10.
Model 06309-11 hose is compatible with Guzzler pump 07090-11.
Model 06309-12 hose is compatible with Guzzler pump 07090-12.