GW Instek GPS-1850D DIGITAL DC POWER SUPPLY 0/18 V
Representative image only.
GW Instek Mfr # GPS-1850D – Item # EW-20017-54
Feature constant voltage and current operation with overload protection
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Wire two single-output models with the same range in a master/slave circuit. A rear panel switch lets you select either master or slave with connections for series or parallel hook-up. Connect in series to double the voltage output; connect in parallel to double the current output. Single-output models also feature a HI/LO range button for current output; the LO range provides a better resolution through a portion of the range.
Triple output model features two independent variable power supplies and one 5 VDC fixed output. Easily set up a master/slave circuit with this model. Connect the two independent power supplies in series or parallel simply by pressing a button.