H-B Instrument B60202-0000 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 70C, 76mm Immersion, Environmentally Friendly

Representative image only.
H-B Instrument Mfr # 602020000 – Item # EW-08008-47
Each thermometer is individually calibrated
  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green”
  • NIST-Certified detailed, laminated report of calibration

PRODUCT OPTIONS

Call For Price

$211.00 - $281.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Length (")12
  • Min Temperature (° C)-10
  • Max Temperature (° C)70
  • Accuracy±0.5°C
  • Division (° C)0.5
  • Thermometer FillBlack Spirit
  • Overall Length (in)12
  • Overall Length (mm)300
  • Immersion Depth (mm)76
  • Stem MaterialYellow-Back Glass
  • Calibration Points0, 37, 56°C
  • DescriptionEasy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 70C, 76mm Immersion, Environmentally Friendly
  • Warranty5 years
5 Year Warranty

Includes

Ring top anti-roll fitting, storage box, and laminated NIST- traceable calibration report

Key Features

  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green”
  • NIST-Certified detailed, laminated report of calibration
  • 5 year warranty

More About this Item

Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. These thermometers are also individually calibrated to NIST standards at specific temperature and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, NIST report of calibration, recyclable packaging, and are RoHS compliant.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat