H-B Instrument B60305-0000 Easy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly

Representative image only.
H-B Instrument Mfr # 603050000 – Item # EW-08008-44
Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green"
  • Statement of accuracy included
  • 5 year warranty

PRODUCT OPTIONS

$29.00USD / Each

$219.50 - $292.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Length (")14
  • Min Temperature (° C)-10
  • Max Temperature (° C)225
  • Accuracy±1.0°C; ±1.5°C above 100°C; ±2.0°C above 200°C
  • Division (° C)1
  • Thermometer FillBlack Spirit
  • Overall Length (in)14
  • Overall Length (mm)350
  • Immersion Depth (mm)350
  • Stem MaterialYellow-Back Glass
  • DescriptionEasy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly
  • Warranty5 years
5 Year Warranty

Key Features

  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green"
  • Statement of accuracy included
  • 5 year warranty

More About this Item

These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications where the use of mercury or other hazardous liquids is not desired. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, statement of accuracy, recyclable packaging, and are RoHS compliant.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat