H-B Instrument B60370-1300 Easy-Read Pocket Liquid-In-Glass Thermometer; -5 to 50C, Aluminum Duplex Case, Environmentally Friendly
Representative image only.
H-B Instrument Mfr # B60370-1300 – Item # EW-90300-35
Rugged case with an eyelet at the top for easy suspension
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")6 19/64
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Accuracy±0.5°C
  • Division (° C)0.5
  • Thermometer FillBlack Spirit
  • Dial / Case MaterialAluminum Duplex
  • Overall Length (in)6.3
  • Overall Length (mm)160
  • Immersion Depth (mm)160
  • Stem MaterialYellow-Back Glass
  • DescriptionEasy-Read Pocket Liquid-In-Glass Thermometer; -5 to 50C, Aluminum Duplex Case, Environmentally Friendly
  • Warranty5 years
5 Year Warranty
Key Features
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty
More About this Item
The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in three case styles; open plastic, aluminum, or nickel-plated brass. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.
Easy-Read® thermometers feature a black liquid and yellow back glass - distinct contrast makes these thermometers very easy to read.