H-B Instrument B60400-0100 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/150C, 50mm Immersion, Environmentally Friendly
Representative image only.
H-B Instrument Mfr # 604000100 – Item # EW-08077-06
PFA coating provides additional safety- no glass or contamination if the thermometer breaks
  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty
PRODUCT OPTIONS
$33.70USD / Each
$219.50 - $292.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")7 29/32
  • Min Temperature (° C)-10
  • Max Temperature (° C)150
  • Accuracy±1.0°C; ±1.5°C above 100°C
  • Division (° C)1
  • Thermometer FillBlack Spirit
  • Overall Length (in)7.9
  • Overall Length (mm)200
  • Immersion Depth (mm)50
  • Stem MaterialPFA-Coated Yellow-Back Glass
  • DescriptionDouble-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/150C, 50mm Immersion, Environmentally Friendly
  • Warranty5 years
5 Year Warranty
Key Features
  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty
More About this Item
Double-safe thermometers are PFA safety coated In the event of breakage the coating will help contain the broken liquid and glass. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, ring top anti-roll fitting, individual serial numbered and have a statement of accuracy.