H-B Instrument B60400-1300 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/150C, Total Immersion, Environmentally Friendly

Representative image only.
H-B Instrument Mfr # B60400-1300 – Item # EW-08077-07
PFA coating provides additional safety- no glass or contamination if the thermometer breaks
  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included

PRODUCT OPTIONS

$21.60USD / Each
$27.00USD / Each

$175.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Length (")7 29/32
  • Min Temperature (° C)-10
  • Max Temperature (° C)150
  • Accuracy±1.0°C; ±1.5°C above 110°C
  • Division (° C)1
  • Thermometer FillBlack Spirit
  • Overall Length (in)7.9
  • Overall Length (mm)200
  • Immersion Depth (mm)200
  • Stem MaterialPFA-coated glass
  • DescriptionDouble -Safe Glass Thermometer, -10/150C, Total, 200 mm, Coated

Key Features

  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included

More About this Item

Double-safe thermometers are PFA safety coated In the event of breakage the coating will help contain the broken liquid and glass. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, ring top anti-roll fitting, individual serial numbered and have a statement of accuracy.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat