H-B Instrument B60509-0200 Enviro-Safe Precision Liquid-In-Glass Thermometer; -1 to 61C, 76mm Immersion, Environmentally Friendly

Representative image only.
H-B Instrument Mfr # 605090200 – Item # EW-08008-78
The only thermometers certified as 100% non-toxic, non-hazardous, biodegradable, and “green” !
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy
  • 5 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Length (\")16 1/2
  • Min Temperature (° C)-1
  • Max Temperature (° C)61
  • Accuracy±1°C; ±0.5°C at 23°C
  • Division (° C)0.1
  • Thermometer FillGreen Spirit
  • Overall Length (in)16.5
  • Overall Length (mm)420
  • Immersion Depth (mm)76
  • Stem MaterialWhite-Back Glass
  • DescriptionEnviro-Safe Precision Liquid-In-Glass Thermometer; -1 to 61C, 76mm Immersion, Environmentally Friendly
  • Warranty5 years
5 Year Warranty

Key Features

  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy
  • 5 year warranty

More About this Item

These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general purpose applications where the use of mercury or other hazardous liquids are not desired. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, anti-roll fitting, individual serial numbered and have a statement of accuracy.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat