H-B Instrument B60570-0800 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 20 to 120F, Closed Plastic Case, Environmentally Friendly

Representative image only.
H-B Instrument Mfr # B60570-0800 – Item # UX-08008-87
Rugged case with an eyelet at the top for easy suspension
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included

PRODUCT OPTIONS

$12.00USD / Each

$185.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Length (")6 19/64
  • Min Temperature (° F)20
  • Max Temperature (° F)120
  • Accuracy±1.0°F
  • Division (° F)1
  • Thermometer FillGreen Spirit
  • Overall Length (in)6.3
  • Overall Length (mm)160
  • Immersion Depth (mm)160
  • Stem MaterialWhite-Back Glass
  • DescriptionEnviro-Safe Liquid-In-Glass Pocket Thermometer; 20 to 120F, Closed Plastic Case, Environmentally Friendly

Key Features

  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included

More About this Item

The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in three case styles; closed metal, open metal, closed plastic, or open plastic. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat