H-B Instrument B61400-3600 Plastic Tube Liquid-in-Glass Thermometer Storage Case; Single, 300mm
Representative image only.
H-B Instrument Mfr # 614003600 – Item # EW-08007-94
Help protect your glass thermometer while in storage
  • 5 year warranty
PRODUCT OPTIONS
$54.00USD / Each
$211.00 - $281.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Length (")12
  • Overall Length (in)12
  • Overall Length (mm)300
  • DescriptionPlastic Tube Liquid-in-Glass Thermometer Storage Case; Single, 300mm
  • Warranty5 years
5 Year Warranty
Key Features
  • 5 year warranty
More About this Item
Clear plastic storage tube holds one glass thermometer and features a twist-lock cap that holds the thermometer securely in place. Choose the size that best fits your thermometer length.