H-B Instrument B61400-9100 Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 254mm
Representative image only.
H-B Instrument Mfr # 614009100 – Item # EW-90311-14
Brass armor protects glass thermometer from damage
  • 3 month warranty
PRODUCT OPTIONS
$29.41USD / Each
$34.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Dial / Case MaterialNickel Plated Brass
  • Overall Length (in)10
  • Overall Length (mm)254
  • Stem MaterialNickel Plated Brass
  • DescriptionNickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 254mm
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Nickel plated brass armor case is designed to protect liquid-in-glass thermometers from accidental breakage during use and storage. The case is available in sizes ranging from 254 mm to 610 mm and features a screw cap with ring and small openings around bulb for circulation.