H-B Instrument B61400-9500 Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 405mm
Representative image only.
H-B Instrument Mfr # 614009500 – Item # EW-90311-16
Brass armor protects glass thermometer from damage
  • 3 month warranty
PRODUCT OPTIONS
$41.65USD / Each
$49.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Dial / Case MaterialNickel Plated Brass
  • Overall Length (in)15.9
  • Overall Length (mm)405
  • Stem MaterialNickel Plated Brass
  • DescriptionNickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 405mm
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Nickel plated brass armor case is designed to protect liquid-in-glass thermometers from accidental breakage during use and storage. The case is available in sizes ranging from 254 mm to 610 mm and features a screw cap with ring and small openings around bulb for circulation.