H-B Instrument B61505-0100 DURAC Thermometer-Hygrometer; 32/248F, 0/100 Percent Humidity Range, Stainless Steel
Representative image only.
H-B Instrument Mfr # B61505-0100 – Item # EW-90311-13
Withstands the elements
  • Stainless steel construction resists corrosion and rust
  • Large 5" (12.7 cm) diameter dial provides easy view of temperature and humidity
  • Hole in top of case allows you to hang in any convenient spot
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Division1°C; 2% RH
  • Accuracy2°F; 4% RH
  • Min Temperature (° C)0
  • Max Temperature (° C)120
  • Description Thermometer-Hygrometer; 32/248F, 0/100 Percent Humidity Range, Stainless Steel
  • Warranty3 month
3 Month Warranty
Key Features
  • Stainless steel construction resists corrosion and rust
  • Large 5" (12.7 cm) diameter dial provides easy view of temperature and humidity
  • Hole in top of case allows you to hang in any convenient spot