Cole-Parmer ASTM 116H Precision, Individually Calibrated 1.250/1.300 Specific Gravity Hydrometer for Heavy Liquids
Representative image only.
Cole-Parmer Item # EW-90311-07
Easily measure specific gravity of petroleum products and heavy liquids
  • Designed to conform to the accuracy requirements of ASTM
  • Temperature standard is 60°F (15.6°C)
  • Individually serialized for easy identification
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")13
  • Hydrometer FormPlain
  • Specific Gravity Range (Min) (%)1.25
  • Specific Gravity Range (Max) (%)1.3
  • Division0.0005
  • Accuracy±0.0005
  • ASTM number116H
  • Length (mm)330
  • Description ASTM 116H Precision, Individually Calibrated 1.250/1.300 Specific Gravity Hydrometer for Heavy Liquids
  • Warranty3 month
3 Month Warranty
Key Features
  • Designed to conform to the accuracy requirements of ASTM
  • Temperature standard is 60°F (15.6°C)
  • Individually serialized for easy identification
  • 3 month warranty
More About this Item
The ASTM precision hydrometers are designed to measure specific gravity of petroleum products and heavy liquids in various ranges from 0.650 to 1.500 with 0.0005 divisions. Available in various calibrated models, these lead free hydrometers are ideal for the food and beverage, chemical, pharmacology, transportation and refrigeration industries.
Equivalent to H-B Instrument 618982500.