Hach 1416100 Nitrate Color Disc Test Kit , 10 ppm Max, 50 Tests/Kit
Representative image only.
Hach Mfr # 1416100 – Item # EW-05545-77
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
PRODUCT OPTIONS
$125.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestNitrate
  • Measurement Points 0.02 to 1 ppm; 0.2 to 10 ppm
  • Reagent PlatformCadmium Reduction
  • Tests Per Kit50
  • DescriptionNitrate Color Disc Test Kit , 10 ppm Max, 50 Tests/Kit
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.