Hach 1421300 Copper, Free Color Disc Test Kit ( 0 to 4 ppm)
Representative image only.
Hach Mfr # 1421300 – Item # EW-05545-86
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
PRODUCT OPTIONS
$114.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestCopper, free
  • Measurement Points0 to 4 ppm
  • Reagent PlatformBicinchoninate
  • Tests Per Kit100
  • DescriptionCopper, Free Color Disc Test Kit ( 0 to 4 ppm)
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.