Hach 145201 Hardness Drop Test Kit , Low Range ( 1 to 20 ppm; 1 to 20gpg)
Representative image only.
Hach Mfr # 145201 – Item # EW-05545-28
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$103.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestHardness, low range
  • Measurement Points1 to 20 ppm; 1 to 20 gpg
  • Reagent PlatformEDTA-powder pillows
  • Tests Per Kit100
  • DescriptionHardness Drop Test Kit , Low Range ( 1 to 20 ppm; 1 to 20gpg)
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field