Hach 145300 Hardness Drop Test Kit , High Range (Range 1 to 30 gpg)
Representative image only.
Hach Mfr # 145300 – Item # EW-05545-30
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$30.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestHardness, high range
  • Measurement Points1 to 30 gpg
  • Reagent PlatformEDTA-bulk powder
  • Tests Per Kit100
  • DescriptionHardness Drop Test Kit , High Range (Range 1 to 30 gpg)
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
This kit does not have a blue case--exception in series.