Hach 146803 Nitrate, High Range Color Disc Test Kit
Representative image only.
Hach Mfr # 146803 – Item # EW-05545-79
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
PRODUCT OPTIONS
$124.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestNitrate, high range
  • Measurement Points1 to 50 ppm
  • Reagent PlatformCadmium Reduction
  • Tests Per Kit100
  • DescriptionNitrate, High Range Color Disc Test Kit
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.