Hach 148002 Sulfite Drop Test Kit , 200 ppm Max, 100 Tests/Kit
Representative image only.
Hach Mfr # 148002 – Item # EW-05545-38
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$168.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestSulfite
  • Measurement Points1 to 20; 10 to 200 ppm
  • Reagent PlatformIodometric
  • Tests Per Kit100
  • DescriptionSulfite Drop Test Kit , 200 ppm Max, 100 Tests/Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field