Hach 18641-00 COD Test Tube Cooling Rack, Holds up to 6 Vials
Representative image only.
Hach Mfr # 18641-00 – Item # EW-99574-54
Optimize water analysis
  • Choose from a variety of accessories
  • 1 year warranty
PRODUCT OPTIONS
$156.00USD / Each
Include Accessories Today!
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Compatible WithHach vials
  • DescriptionCOD Test Tube Cooling Rack, Holds up to 6 Vials
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Stainless steel rack for holding hot COD vials or test tubes prior to reading. Fit Hach COD vials and Test 'N Tube vials.