Hach 201002 Cyn-3 Color Disc Test Kit , Cyanide; 100 Tests/Kit
Representative image only.
Hach Mfr # 201002 – Item # EW-05545-90
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$310.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestCyanide
  • Measurement Points0.01 to 0.3 ppm
  • Reagent PlatformPyridine-Pyrazalone
  • Tests Per Kit100
  • DescriptionCyn-3 Color Disc Test Kit , Cyanide; 100 Tests/Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.