Hach 2113301 PN-10 Phosphonate Color Disc Test Kit without UV Lamp and Power Supply
Representative image only.
Hach Mfr # 2113301 – Item # EW-05545-52
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • No UV lamp or power supply included, must purchase separately (see 05545-63 for kit with UV lamp and power supply)
  • 100 tests included
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • TestPhosponate
  • Measurement Points1 to 5 ppm
  • Reagent PlatformUV Photolysis
  • Tests Per Kit100
  • DescriptionPhosphonate Color Disc Test Kit without UV Lamp and Power Supply
  • Warranty1 year
1 Year Warranty
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • No UV lamp or power supply included, must purchase separately (see 05545-63 for kit with UV lamp and power supply)
  • 100 tests included
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
Phosphonate Test Kit, Model PN-10, without UV Lamp and Power Supply