Hach 2182000 Nitrogen Color Disc Test Kit , 0.5 ppm Max, 100 Tests/Kit
Representative image only.
Hach Mfr # 2182000 – Item # EW-05545-85
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
PRODUCT OPTIONS
$94.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Test Nitrogen, Low range
  • Measurement Points 0.01 to 0.50 ppm
  • Tests Per Kit100
  • DescriptionNitrogen Color Disc Test Kit , 0.5 ppm Max, 100 Tests/Kit
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.