Hach 2194100 Copper, Free and Total Color Disc Test Kit
Representative image only.
Hach Mfr # 2194100 – Item # EW-05545-88
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$129.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestCopper, free and total
  • Measurement Points0 to 4 ppm
  • Reagent PlatformBicinchoninate
  • Tests Per Kit150
  • DescriptionCopper, Free and Total Color Disc Test Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.