Hach 222800 Chromium, Hexavalent Color Disc Test Kit
Representative image only.
Hach Mfr # 222800 – Item # EW-05545-82
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • TestChromium, hexavalent
  • Measurement Points0.1 to 1.5 ppm
  • Reagent PlatformDiphenylcarbazide, Hypobromite Oxidation
  • Tests Per Kit50
  • DescriptionChromium, Hexavalent Color Disc Test Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.