Hach 223400 Color Disc Test Kit , Pt-Co Scale, 500 Units Max
Representative image only.
Hach Mfr # 223400 – Item # EW-05545-84
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$140.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestColor
  • Measurement Points0 to 100 units; 0 to 500 units
  • Reagent PlatformAPHA Platinum-Cobalt
  • DescriptionColor Disc Test Kit , Pt-Co Scale, 500 Units Max
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.