Hach 223801 Hydrogen Sulfide Color Disc Test Kit
Representative image only.
Hach Mfr # 223801 – Item # EW-05545-98
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$322.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestHydrogen sulfide, sulfide
  • Measurement Points 0.01 to 0.55 ppm; 0.05 to 2.25 ppm; 0.25 to 10 ppm
  • Reagent PlatformHydrogen Sulfide Methylene Blue
  • DescriptionHydrogen Sulfide Color Disc Test Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.