Hach 224902 PHosPHorous Color Disc Test Kit (0.1 to 5; 1 to 50 ppm)
Representative image only.
Hach Mfr # 224902 – Item # EW-05545-41
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$223.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestPhosphorous
  • Measurement Points0.1 to 5 ppm; 1 to 50 ppm
  • Tests Per Kit100
  • DescriptionPHosPHorous Color Disc Test Kit (0.1 to 5; 1 to 50 ppm)
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.