Hach 2314500 Alkalinity Drop Test Kit , High Level
Representative image only.
Hach Mfr # 2314500 – Item # EW-05545-02
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$77.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestAlkalinity, high range
  • Measurement Points23 to 495 gpg; 385 to 8,500mg/L
  • Reagent PlatformSulfuric Acid
  • Tests Per Kit100
  • DescriptionAlkalinity Drop Test Kit , High Level
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field