Hach 2515050 Dissolved Oxygen Color Disc Test Kit
Representative image only.
Hach Mfr # 2515050 – Item # EW-05545-94
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$214.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestDissolved oxygen
  • Measurement Points0 to 15 ppm
  • Reagent PlatformHRDO
  • Tests Per Kit25
  • DescriptionDissolved Oxygen Color Disc Test Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.