Hach 2518050 Ozone, High Range Color Disc Test Kit
Representative image only.
Hach Mfr # 2518050 – Item # EW-05545-87
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$197.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestOzone, High range
  • Measurement Points0.05 to 1.5 ppm
  • Reagent PlatformIndigo Trisulfonate
  • Tests Per Kit25
  • DescriptionOzone, High Range Color Disc Test Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.