Hach 255600 Iron, Low Range Color Disc Test Kit
Representative image only.
Hach Mfr # 255600 – Item # EW-05545-64
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
PRODUCT OPTIONS
$152.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestIron, low range
  • Measurement Points 0 to 0.2 ppm; 0 to 1.0 ppm
  • Reagent PlatformFerroZine
  • Tests Per Kit100
  • DescriptionIron, Low Range Color Disc Test Kit
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.