Hach 2744940 Chloride Test Strips Low Range , 40 (Range 30 to 600 mg/L)
Representative image only.
Hach Mfr # 2744940 – Item # EW-05546-16
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$80.45USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • TestChloride, low range
  • Measurement Points30 to 600 ppm
  • Reagent PlatformStrips
  • Tests Per Kit40
  • DescriptionChloride Test Strips Low Range , 40 (Range 30 to 600 mg/L)
ISO9001 Certified Supplier
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test chloride as Cl- from 30 to 600 mg/L. 40 test strips included.