Hach 2755250 5 In 1 Water Quality Test Strips, 50
Representative image only.
Hach Mfr # 2755250 – Item # EW-05546-02
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$29.65USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestpH, free and total chlorine, hardness, alkalinity
  • Reagent PlatformStrips
  • Tests Per Kit50
  • Description5 In 1 Water Quality Test Strips, 50
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
The strips will test free and total chlorine from 0 to 10 mg/L; pH from 6.8 to 8.4; total hardness as CaCO3 from0 to 25 gpg; and total alkalinity as CaCO3 from 0 to 425 mg/L.
50 tests are included.