Hach 2757150 PHosPHorous/O-Phosphate Test Strips, 50
Representative image only.
Hach Mfr # 2757150 – Item # EW-05546-44
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$34.25USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • TestPhosphorus, o-phosphate
  • Measurement Points0, 5, 15, 30, 50 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionPHosPHorous/O-Phosphate Test Strips, 50
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests Phosphorus and o-Phosphate (reactive)(PO4) from 0 to 50 mg/L. 50 test strips included.