Hach 27603-45 TOC Test Kit, Low Range, 0 to 20 mg/L
Representative image only.
Hach Mfr # 27603-45 – Item # EW-99574-28
Optimize water analysis
  • Choose from a variety of accessories
PRODUCT OPTIONS
$681.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithHACH DR/870
  • DescriptionTOC Test Kit, Low Range, 0 to 20 mg/L
Key Features
  • Choose from a variety of accessories
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Kits contain leak-proof vials. Caps provide a positive seal that traps volatile material and ensures complete digestion. Reagents are premixed, minimizing dangerous spills and handling of corrosive and toxic chemicals. Small reagent volumes mean less silver and mercury waste. Pack of 25 reagent vials.