Hach 27604-45 TOC Test Kit, High Range, 20 to 700 mg/L
Representative image only.
Hach Mfr # 27604-45 – Item # EW-99574-29
Optimize water analysis
  • Choose from a variety of accessories
  • 1 year warranty
PRODUCT OPTIONS
$746.00USD / Each
Include Accessories Today!
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithHACH DR/870
  • DescriptionTOC Test Kit, High Range, 20 to 700 mg/L
  • Warranty1 year
1 Year WarrantyCanadian Standards AssociationEPA United States Environmental protection agencyUL Listed.
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Replacement electrodes and probes, sample cells, carrying cases, and much more!
Kits contain leak-proof vials. Caps provide a positive seal that traps volatile material and ensures complete digestion. Reagents are premixed, minimizing dangerous spills and handling of corrosive and toxic chemicals. Small reagent volumes mean less silver and mercury waste. Pack of 25 reagent vials.