Hach 2890200 Free Chlorine High Range Test Strips, 100
Representative image only.
Hach Mfr # 2890200 – Item # EW-05546-24
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$34.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestChlorine, free; high range
  • Measurement Points0, 25, 50, 100, 200, 400, 600 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionFree Chlorine High Range Test Strips, 100
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test free chlorine as Cl2 at high level from 0 to 600 mg/L. 100 test strips included.