Hach AMMONIA STRIPS Ammonia (Nitrogen) Test Strips, 0 to 6.0 mg/L; 25 Strips/Pk
Representative image only.
Hach Mfr # 2755325 – Item # EW-05546-06
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$38.55USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestAmmonia, nitrogen
  • Measurement Points0, 0.25, 0.5, 1, 3, 6 pm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionAmmonia (Nitrogen) Test Strips, 0 to 6.0 mg/L; 25 Strips/Pk
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests Nitrogen Ammonia NH3-N from 0 to 6 mg/L. 25 test strips included.