Hach DR1900 Portable Water Spectrophometer

Representative image only.
Hach Mfr # DR1900-01H – Item # EW-99573-20
Designed for drinking water and wastewater method analysis
  • Wavelength range of 340 to 800 nm includes most water testing methods
  • Select from pre-programmed water quality methods or user-customizable methods
  • 500 GLP compliant data sets with USB connectivity
  • IP67-rated for waterproofing
  • Operates on four AA batteries for field use, or optional AC adapter for laboratory

$4,913.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Light SourceXenon flash
  • Wavelength(s) (nm)340 to 800
  • Min Operating Temperature (° F)50
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)10
  • Max Operating Temperature (° C)40
  • WaterproofYes
  • Data LoggingYes
  • Memory Size500 measurements
  • Length (in)7
  • Width (in)4
  • Height (in)10 1/2
  • Description Portable Water Spectrophometer
  • Warranty1 year
Conformite EuropeenneIP 671 Year Warranty

Includes

four sample cell adapters, 10 mL matching pair of sample cells, sample-light cover, dust cover, and four AA batteries.

Key Features

  • Wavelength range of 340 to 800 nm includes most water testing methods
  • Select from pre-programmed water quality methods or user-customizable methods
  • 500 GLP compliant data sets with USB connectivity
  • IP67-rated for waterproofing
  • Operates on four AA batteries for field use, or optional AC adapter for laboratory
  • 1 year warranty

More About this Item

Accuracy and dependability in a portable meter designed for routine visible (340 to 800 nm) water and wastewater methods. The Hach and Hach/Lange preprogrammed and user-customizible water quality methods offer absorbance, % transmittance, and concentration measurements. Select from user programs, favorites, single wavelegth, multiple wavelength, and time course modes.
Collect up to 500 data sets with GLP compliance with USB mini connecticity. A range of sample cells are available (not included): 10 x 10 mm, 1-in rectangular, 13 mm, 16 mm, 1-in round, 1 cm/10 mL, and flow-thru cell.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat